Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Navigator™ XL Balance

Ultrafast stabilization in less than one second provides fast, accurate and repeatable weighing results

Manufacturer:  OHAUS™ 83033074

 View more versions of this product

Catalog No. 01-918-315


Due to product restrictions, please to purchase or view availability for this product.


Description

Description

Two touchless sensors free up your hands for handling samples and limiting keypad wear and tear.
 
  • Superior mechanical overload protection system and integrated shipping lock
  • Withstands loads up to 400% of its rated capacity
  • Optional RS-232 or USB or ethernet interface kits with integrated cables
  • Up front illuminated level indicator
  • Removable stainless-steel weighing platform
  • Menu lockswitch
  • Integral security bracket and integral shipping lock
  • Stability indicator, overload and underload indicators, low battery indicator, auto shut-off
  • ABS plastic housing; 304 stainless steel pan
Specifications

Specifications

10 kg
0.25 A
  • Electromagnetic Compatibility: C-Tick, CE Class B, FCC Class B, ICES-003 Class B
  • Safety (AC adaptors only): C-Tick, CCC, CE, cCSAus, KTL, PSE, TUV-GS; Compliant to Directive 2002/96/EC on waste electrical and electronic equipment (WEEE); Compliant to Directive 2002/95/EC on the restriction of the use of certain hazardous substances in electrical and electronic equipment (RoHS)
  • Quality: OHAUS quality management system registered to ISO 9001:2008
28.2cm
1 g
7.4cm
AC adapter or internal rechargeable battery
1g
ABS Plastic, Stainless Steel
50 to 104°F
100/240V
4.3 lb.
20.4cm
Pound, ounce (decimal/fractional), pound: ounce (d
Backlit LCD
Navigator™ XL scale
RS-232 or USB connectivity
4 Mechanical keys, raised and with tactile feedbac
11.1 in.
2.9 in.
50/60Hz
±2g
NVL10000/1
≤1 sec.
10 to 40°C
2kg
8 in.
100/240V, 50/60Hz
2 Years
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit