Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OHAUS School Balance
Ideal teaching tool to introduce students to precision mass measurement. OHAUS School Balances measure small items such as paper clips, marbles and coins, as well as heavier objects such as books.
$67.50
Specifications
Capacity | 4.4 lb., 2 kg |
---|---|
Warranty | 1 Year |
Alert:
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Catalog Number | Mfr. No. | Depth (English) Pan | Depth (Metric) Pan | Includes | Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Depth (English) Pan | Depth (Metric) Pan | Includes | Type | Price | Quantity & Availability | ||
S63485
![]() |
OHAUS
80000029 |
1.25 in. | 3.2 cm | 8 Piece Metal mass set, bowl type pans for use in | School Balance |
Each for $67.50
|
|
Description
- Dual pointers allow students to view results from either side of the balance
- Interchangeable square pans remove for easy pouring, cleaning and storing
- Pivoting beam provides accuracy to 0.5g
- Capacity: 70.5 oz. (2000g)
- Includes 18-piece plastic weight set
School Balance Measurement Bundle
- Includes six Ohaus school balances (S01976) and one measurement skills assessment kit (S97292)
Warranty and Services
One year
Specifications
4.4 lb., 2 kg | |
1 Year |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title