Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Spring Scales: Demonstration Dial Type Products for Science Education

Designed to stand up to heavy student use and withstand sudden load releases without breaking.

Manufacturer:  OHAUS™ 80000050

 View more versions of this product

Catalog No. S40136


Add to cart

Description

Description

  • Simple zero adjustment
  • Easy-to-read black graduations on a sturdy frosted-aluminum background
  • Color-coded styles for easy identification of capacity

Order Info

Dial- and pull-type scales are available as well as 8 in. (20.3cm) class demonstration size scales.

Specifications

Specifications

External
8 in.
User manual
8015-00
11 in.
4.4 lb., 2 kg
20.32 cm
Plastic, Metal
Dial Spring Scale
8 cm
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit