Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Starter Water Analysis Bench Meters: Model 2100
SCI_ED

Durable, accurate bench meters for a variety of lab applications. OHAUS Starter Water Analysis Bench Meters: Model 2100 provides reliable pH testing with intuitive software/organized display for usability.

Specifications

Display Type LCD
Resolution 0.01 pH; 1 mV; 0.1°C
View More Specs

Includes: Water analysis meter; available with or without electrode

Products 2
Catalog Number Mfr. No. Includes Model Price Quantity & Availability  
Catalog Number Mfr. No. Includes Model Price Quantity & Availability  
S05073
SCI_ED
View Documents
OHAUS™
30057495/EMD
Water analysis meter with ST210 2-in-1 plastic refillable pH electrode and pH buffer powder sachet ST2100-E This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
S05074
SCI_ED
View Documents
OHAUS™
30057496/EMD
Water analysis meter with ST210 2-in-1 plastic refillable pH electrode, STTEMP30 temperature electrode, and pH buffer powder sachet ST2100-F This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description

Description

  • Clearly marked input keys
  • Built-in or standalone electrode holders
  • Reliable calibration for accurate measurement
  • Built-in electrode arm for simple and convenient operation
  • Auto buffer minimizes errors during calibration process
  • Reliable measurements
  • Cost effective
  • Size: 220 x 175 x 78mm
  • Weight: 0.75kg
Specifications

Specifications

LCD
0.01 pH; 1 mV; 0.1°C
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit