Learn More
OHAUS™ Starter Water Analysis Portable Meters Products for Science Education
Portable, lightweight meters feature productivity and versatility in the field or lab. OHAUS Starter Water Analysis Portable Meter is a compact meter with intuitive software/organized display for ultimate usability.
$106.44 - $636.00
Save up to 46%
Specifications
Battery Life 250 hr.
Battery Type Four AAA
Data Points 30
Display Type LCD
IP Rating 54
View More Specs
Products 3
Catalog Number Mfr. No. Calibration Points Includes Inputs Temperature Range (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Calibration Points Includes Inputs Temperature Range (Metric) Price Quantity & Availability  
S05081 Products for Science Education View Documents OHAUS™
83033964
1 Point, 3 pre defined standard Water analysis meter, STCON3 four-pole, conductivity electrode (70 μS/cm-200ms/cm), IP54 sets (1413 μS/cm and 12.88ms/cm), four AAA batteries Mini-DIN 0° to 100°C Each for $463.00
Add to cart
S05083 Products for Science Education View Documents OHAUS™
30031655
1 or 2 Points, 100% and 0% Water analysis meter, IP54 sets, electrode clip, wrist strap and 4 AAA batteries, STDO11 Galvanic DO electrode and STTEMP30 temperature electrode BNC, Cinch, NTC 30kilohm 0° to 50°C Each for $636.00
Add to cart
S05082 Products for Science Education View Documents OHAUS™
30031656
1 or 2 Points, 100% and 0% Water analysis meter, IP54 sets, electrode clip, wrist strap and 4 AAA batteries BNC, Cinch, NTC 30kilohm 0° to 50°C Each for $106.44
Save 46% Reg : $198.01
Add to cart
Description
Description
  • IP54 protection shields water, dust damage
  • Compact, ergonomic design
  • Reliable calibration for accurate measurement
  • 30-set memory storage
  • Quick calibration data recall
Starter 300C Conductivity Meter
  • Four-pole linear electrode safeguards polarization/pollution effects
  • Automatic temperature compensation with an adjustable temperature coefficient
  • Accurate measurement of conductivity and TDS levels
  • Wide variety of applications
  • Withstands extreme conditions
  • Includes 4-pole conductivity electrode
Starter 300D Dissolved Oxygen Meter
  • Accurate, reliable measurement critical to aquatic life and water quality
  • Icon display confirms accuracy of calibration process
  • Push-button toggling between ppm, mg/L, percentage
  • Galvanic electrode ready immediately after powering
  • Model ST300D-B is for meter only
  • Model ST300D includes a Galvanic DO Electrode
Specifications
Specifications
250 hr.
30
54
0.1°C
Four AAA
LCD
Four AAA