Learn More
OHAUS™ Starter Water Analysis Portable Meters
SCI_ED
Portable, lightweight meters feature productivity and versatility in the field or lab. OHAUS Starter Water Analysis Portable Meter is a compact meter with intuitive software/organized display for ultimate usability.
Supplier:  OHAUS™ 30031656/EMD
Catalog No. S05082
Add to Cart
Description
Description
  • IP54 protection shields water, dust damage
  • Compact, ergonomic design
  • Reliable calibration for accurate measurement
  • 30-set memory storage
  • Quick calibration data recall
Starter 300D Dissolved Oxygen Meter
  • Accurate, reliable measurement critical to aquatic life and water quality
  • Icon display confirms accuracy of calibration process
  • Push-button toggling between ppm, mg/L, percentage
  • Galvanic electrode ready immediately after powering
  • Model ST300D-B is for meter only
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Water analysis meter, IP54 sets, electrode clip, wrist strap and 4 AAA batteries
Automatic or Manual
0.1°C
30
IP54
Four AAA
250 hr.
1 or 2 Points, 100% and 0%
0°C to 50°C
LCD
BNC, Cinch, NTC 30kilohm
0.1%; 1% 0.1 mg/L; 1mg/L 0.01 ppm; 0.1 ppm 0.1°C
4 x AAA
Product Suggestions
Product Suggestions