Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Valor™1000 Compact Scale

Economical, multi-purpose scale with standard features. OHAUS Valor 1000 Compact Scales include a stainless weighing platform, ABS plastic housing with leveling adjustment, and internal rechargeable battery.

Manufacturer:  OHAUS™ 83998128

 View more versions of this product

Catalog No. 02-112-102


Due to product restrictions, please to purchase or view availability for this product.


Description

Description

  • Large, removable stainless steel weighing pan
  • Standard software includes checkweighing mode, accumulation mode and multiple units of measure
  • Adjustable leveling feet and front panel level bubble
  • Mid profile ABS plastic housing
  • AC adaptor (supplied) or internal rechargeable battery
  • LCD with blue backlight
Specifications

Specifications

0.005kg
32 to 104°F
120V
3.2kg
10 in.
25cm
Valor™ 1000 compact scale
80-100 hr.
Digital external span calibration
7.1 in.
18cm
LCD
4.3 in.
AC adapter, leveling adjustment and front panel level bubble
V11P15
<3 sec.
0 to 40°C
7 lb.
9.9 in.
25cm
15 kg
4 Mechanical switch
Internal rechargeable Lead Acid battery
ISO 9001:2008, NSF, USDA-AMS, HACCP, CE, AS/NZS4251.1 Emission/AS/NZS4252.1 Immunity, FCC Class A Part 15 operation, Industry Canada Class A, cULus
12 in.
30.7cm
Gram, kilogram, ounce, pound
11cm
ABS Plastic, Stainless Steel
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit