Learn More
OHAUS™ Valor™ 3000 Compact Food Scales
Stainless-steel design in a high-performance, easy-to-use, portable scale
Manufacturer:  OHAUS™ 83998177
 View more versions of this product
Catalog No. 02-115-332
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Software for weighing, percent weighing and specific gravity determination
  • All stainless-steel construction
  • Compact space-saving footprint
  • AC adapter (supplied) or 4 C batteries
  • Adjustable levelling feet and bubble (only for XW models)
  • Load cell lock, user adjustable filtering and optional food accessories
  • LCD with white LED backlight
Certifications
NSF Certified
Compliance
Supports HACCP compliance; washable models available.
Specifications
Specifications
Valor™ 3000 compact food scale
50 hr.
Digital external span calibration
8.5 x 7.3 x 3 in. (21.8 x 18.6 x 7.7cm)
100/240V 50/60Hz
3 in.
50/60Hz
5.8 in.
14.6cm
Stainless Steel
0.1g
32 to 104°F
100/240V
1.3kg
7.3 in.
18.6cm
3 Mechanical switch
4 C
ISO 9001:2008, NSF/ANSI 169-2005, USDA/AMS, HACCP, CE, AS/NZS4251.1 Emission/AS/NZS4252.1 Immunity, FCC Class B Part 15 operation, Industry Canada Class A, cULus AC power pack, Directive 2002/96/EC, NTEP CoC No. 08-008
LCD
Gram, kilogram, ounce, pound, fluid ounce
7.7cm
AC adapter, adjustable leveling feet and bubble, load cell lock
8.5 in.
21.8cm
V31XH4
<3 sec.
0 to 40°C
2.9 lb.
6.2 in.
15.8cm
4 kg