Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Valor™ 3000 Xtreme Portable Scale

Compact portioning scale offers high precision and extreme durability for the harshest environments

Manufacturer:  OHAUS™ 83998134

 View more versions of this product

Catalog No. 02-115-550


Due to product restrictions, please to purchase or view availability for this product.


Includes: AC adapter, load cell lock

Description

Description

High Precision NSF certified and listed scale with USDA/AMS accepted housing features the accuracy of the Scout™ Pro with precision up to 20,000d. User-selectable filtering, auto-zero tracking and stability settings enable use in drafty, unstable environments.
 
  • Full T304 stainless-steel housing, removable stainless-steel weighing pan
  • Corrosion-resistant keyboard, great for areas where harsh chemicals are used
  • SmarText™ user interface utilizes bright, backlit LCD plus simple text prompts
  • Overload protection rated at 10x capacity
  • Percentage weighing mode can automatically compute Baker's Percentages
  • Specific Gravity Mode offers rapid, easy determination without need for manual calculations—great for checking density, aeration of mixes
  • Durable, easy to clean
  • Adjustable rubber feet
  • Waterproof models (V31XW3, V31XW6) feature full NEMA 4X/IP65 protection and up-front level bubble and NTEP certified models available
 
Applications
Laboratory, industrial, food service, baking

Service Plan

Service agreement available.

Certifications

NSF Meets requirements for HACCP conformity, USDA/AMS, UL and CE, NTEP certified models V31XW3, V31XW6

Compliance

EMC Approvals: FCC Class B, Industry Canada, CE.

Specifications

Specifications

Valor™ 3000 Xtreme portable food portioning scale
3 sec.
0 to 40°C
3.1 lb.
6.2 in.
15.8cm
0.6 lb., 300 g
50 hr.
Digital external span calibration
8.5 x 7.3 x 3 in. (21.8 x 18.6 x 7.7cm)
100/120V 50/60Hz
3 in.
50/60Hz
5.8 in.
14.6cm
Stainless Steel
0.2g
32 to 104°F
100/120V
1.4kg
7.3 in.
18.6cm
3 Mechanical switch
4 C
ISO 9001:2008, NSF, USDA/AMS, CE, AS/NZS4251.1 Emission/AS/NZS4252.1 Immunity, FCC Class B Part 15 operation, Industry Canada Class A, cULus, NTEP CoC No. 08-008
LCD
Gram, kilogram, pound, pound: ounce (fractional), fluid ounce (Water)
7.7cm
AC adapter, adjustable leveling feet and bubble, load cell lock
8.5 in.
21.8cm
V31XW301 (Waterproof)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit