Learn More
Azzota Corp OICR-9429 DFS Item
Manufacturer:  Azzota Corp C1243
OICR-9429 is a potent and selective chemical probe suitable to help dissect the biological role of WDR5 (Kdisp < 100 nM). OICR-9429 selectively inhibited proliferation and induced differentiation in p30-expressing human AML cells.
Catalog No. 50-161-3157