Learn More
Cole-Parmer Ultrafilter UFS-SUPERPLUS 5 Oil/Water Separator, 0.5 GPH  DFS Item
Manufacturer:  Cole-Parmer 2957800
Note: Not recommended for use with emulsified condensates.
Catalog No. 50-628-620