Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OK-1™ Full Finger Anti-Vibration Gloves
Click to view available options
Size:
Large
X-Large
Description
- Premium grain leather with stretch nylon back
- Designed to form to the natural curve of the hand for increased comfort
- Machine washable
Specifications
Color | Brown |
Hand | Specific |
Material | Grain Cowhide Leather |
Product Type | Work Gloves |
Size | X-Large |
For Use With (Application) | Construction, Machine Operation, and Manufacturing |
Glove Cuff Style | Open |
Series | OK990 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction