Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Okadaic acid sodium salt, 98%

Click to view available options
Quantity:
50μg
100μg
300μg
209266-80-8
C44H67NaO13
826.997
MFCD00210208
BYHIFOCTDVNQQT-GHIYGBLASA-M
okadaic acid sodium salt,okadaic acid sodium salt from prorocentrum concavum,okadaic acid sodium salt from prorocentrum concavum hplc , film
71308608
sodium;(2R)-3-[(2S,6R,8S,11R)-2-[(E,2R)-4-[(2S,2'R,4R,4aS,6R,8aR)-4-hydroxy-2-[(1S,3S)-1-hydroxy-3-[(2S,3R,6S)-3-methyl-1,7-dioxaspiro[5.5]undecan-2-yl]butyl]-3-methylidenespiro[4a,7,8,8a-tetrahydro-4H-pyrano[3,2-b]pyran-6,5'-oxolane]-2'-yl]but-3-en-2-yl]
CC1CCC2(CCCCO2)OC1C(C)CC(C3C(=C)C(C4C(O3)CCC5(O4)CCC(O5)C=CC(C)C6CC(=CC7(O6)C(CCC(O7)CC(C)(C(=O)[O-])O)O)C)O)O.[Na+]
209266-80-8
C44H67NaO13
826.997
MFCD00210208
BYHIFOCTDVNQQT-GHIYGBLASA-M
okadaic acid sodium salt,okadaic acid sodium salt from prorocentrum concavum,okadaic acid sodium salt from prorocentrum concavum hplc , film
71308608
sodium;(2R)-3-[(2S,6R,8S,11R)-2-[(E,2R)-4-[(2S,2'R,4R,4aS,6R,8aR)-4-hydroxy-2-[(1S,3S)-1-hydroxy-3-[(2S,3R,6S)-3-methyl-1,7-dioxaspiro[5.5]undecan-2-yl]butyl]-3-methylidenespiro[4a,7,8,8a-tetrahydro-4H-pyrano[3,2-b]pyran-6,5'-oxolane]-2'-yl]but-3-en-2-yl]
CC1CCC2(CCCCO2)OC1C(C)CC(C3C(=C)C(C4C(O3)CCC5(O4)CCC(O5)C=CC(C)C6CC(=CC7(O6)C(CCC(O7)CC(C)(C(=O)[O-])O)O)C)O)O.[Na+]
  • Potent inhibitor of serine/threonine-specific protein phosphatases 1 and 2A (PP1 and PP2A)
  • Does not inhibit tyrosine or alkaline phosphatases
UN Number UN3462
Solubility Information Soluble in DMSO or ethanol
Formula Weight 827
Physical Form Powder
Percent Purity 98%
Color White
Quantity 50μg
Chemical Name or Material Okadaic acid sodium salt

GHS H Statement
H300-H310-H330-H315
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: I; Proper Shipping Name: TOXINS, EXTRACTED FROM LIVING SOURCES, SOLID, N.O.S.

storageNote1 : -30° to -10°C