Learn More
Abnova™ OKL38 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00029948T01
Catalog No. 89-954-993
Add to cart
Specifications
Specifications
293T
NM_182980.1
pCMV-OKL38 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
OKL38 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 52.58kDa
Transient overexpression cell lysate was tested with anti-OKL38 antibody (Mfr. No. H00029948-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human