Alfa Aesar™ Oleic acid, 99%

Manufacturer: Alfa Aesar™ 3199706

Quantity 5g

 View more versions of this product

Catalog No. AA3199706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 112-80-1
    Formula Weight 282.47
    MDL Number MFCD00064242
    Beilstein 1726542
    Merck Index 14,6828
    Synonym cis-9-Octadecenoic acid
    Chemical Name or Material Oleic acid

    Specifications

    Formula Weight 282.47
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see ... on this label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.
    Density 0.887g/mL
    Physical Form Liquid
    Quantity 5g
    Synonym cis-9-Octadecenoic acid
    Refractive Index 1.4610
    Color Colorless to pale yellow
    Sensitivity Air Sensitive
    MDL Number MFCD00064242
    Beilstein 1726542
    Boiling Point 286°C (100mmHg)
    Melting Point 13° to 14°C
    TSCA TSCA
    CAS 112-80-1
    Merck Index 14,6828
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    RTECS Number RG2275000
    Chemical Name or Material Oleic acid
    Assay 99%

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see ... on this label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit