Learn More
Alfa Aesar™ Oleoyl-L-alpha-lysophosphatidic acid sodium salt
Manufacturer:  Alfa Aesar™ J66836LB0
 View more versions of this product
Catalog No. AAJ66836LB0
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
5mg
-30°C to -10°C
Oleoyl-L-alpha-lysophosphatidic acid sodium salt
C21H40NaO7P
1-O-9Z-Octadecenoyl-sn-glyceryl-3-phosphoric acid sodium salt; Lysophosphatidic Acid
Soluble in water up to 5mg/ml with sonication
White to off-white
Waxy solid or powder
22556-62-3
458.5
Light and Air Sensitive and Hygroscopic