Alfa Aesar™ Oleyl alcohol, 99+%

Manufacturer: Alfa Aesar™ H3347514

Quantity 25g

 View more versions of this product

Catalog No. AAH3347514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 143-28-2
    Molecular Formula C18H36O
    Formula Weight 268.48
    MDL Number MFCD00002993
    Beilstein 1723962
    Synonym cis-9-Octadecen-1-ol
    Chemical Name or Material Oleyl alcohol

    Specifications

    Formula Weight 268.48
    Molecular Formula C18H36O
    MDL Number MFCD00002993
    Beilstein 1723962
    Health Hazard 1 Warning
    Boiling Point 206° to 207°C (13mmHg)
    Melting Point ≈6°C
    Flash Point 180°C (356°F)
    Quantity 25g
    Recommended Storage −30° to −10°C
    RTECS Number RG4120000
    TSCA TSCA
    CAS 143-28-2
    Synonym cis-9-Octadecen-1-ol
    Health Hazard 2 GHS H Statement
    H315
    Causes skin irritation.
    Health Hazard 3 GHS P Statement
    P280-P264-P362-P321-P332+P313-P302+P352
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    IF ON SKIN: Wash with plenty of soap and water.
    Chemical Name or Material Oleyl alcohol
    Assay ≥99%

    Description

    Applications
    Oleyl alcohol is used in softening and lubrication of textile fabrics, in the production of carbon paper, stencil paper, and printing ink. It finds application in cosmetic products viz skin creams, lotions as a thickner, and in hair conditioners and hair coating shampoos. It is utilized as an antifoaming agent and cutting lubricant, as the precursor for the preparation of its sulfuric ester derivatives, which are used in detergents and wetting agents. It plays a vital role in various formulations for drug delivery.

    Solubility
    Miscible with alcohol and ether. Slightly miscible with carbon tetrachloride. Immiscible with water.

    Notes
    Incompatible with oxidizing agents and strong acids.

    GHS H Statement
    H315
    Causes skin irritation.

    −30° to −10°C

    GHS P Statement
    P280-P264-P362-P321-P332+P313-P302+P352
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    IF ON SKIN: Wash with plenty of soap and water.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit