Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oleyl alcohol, ca. 60%, technical, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
25L
Packaging:
Glass Bottle
Metal drum
143-28-2
C18H36O
268.47
MFCD00002993
ALSTYHKOOCGGFT-KTKRTIGZSA-N
oleyl alcohol,cis-9-octadecen-1-ol,z-octadec-9-en-1-ol,ocenol,conditioner 1,dermaffine,lancol,novol,oceol,oleol
5284499
CHEBI:73504
(Z)-octadec-9-en-1-ol
CCCCCCCCC=CCCCCCCCCO
143-28-2
C18H36O
268.47
MFCD00002993
ALSTYHKOOCGGFT-KTKRTIGZSA-N
oleyl alcohol,cis-9-octadecen-1-ol,z-octadec-9-en-1-ol,ocenol,conditioner 1,dermaffine,lancol,novol,oceol,oleol
5284499
CHEBI:73504
(Z)-octadec-9-en-1-ol
CCCCCCCCC=CCCCCCCCCO
Infrared Spectrum Authentic
Density 0.8400g/mL
Quantity 25L
Melting Point 5.0°C
Packaging Metal drum
Flash Point 170°C
Specific Gravity 0.84
Refractive Index 1.4590 to 1.4640
Boiling Point 207.0°C (13.0 mmHg)
Color Colorless to Yellow
Assay Percent Range ca. 60%
Linear Formula CH3(CH2)7CH=CH(CH2)8OH
Beilstein 01, 453
Merck Index 15, 6924
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alcohol and ether, soluble in warbontetrachloride
Formula Weight 268.47
Physical Form Viscous Liquid
Percent Purity ca. 60%
Grade Technical
Chemical Name or Material Oleyl alcohol
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.