Oleyl alcohol, tech. 85%, Alfa Aesar

Manufacturer: Alfa Aesar A1801836

Quantity 500g

 View more versions of this product

Catalog No. AAA1801836

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 143-28-2
    Physical Form Liquid
    Assay Percent Range 0.85
    Molecular Formula CH3(CH2)7CH=CH(CH2)7CH2OH
    Formula Weight 268.49
    Chemical Name or Material Oleyl alcohol
    Grade Technical Grade

    Specifications

    Formula Weight 268.49
    Quantity 500g
    Physical Form Liquid
    Density 0.849
    Assay Percent Range 0.85
    Molecular Formula CH3(CH2)7CH=CH(CH2)7CH2OH
    CAS 143-28-2
    Chemical Name or Material Oleyl alcohol
    Grade Technical Grade

    Description

    Applications
    Oleyl alcohol is used in softening and lubrication of textile fabrics, and in the production of carbon paper, stencil paper, and printing ink. It finds application in cosmetic products viz skin creams and lotions as a thickner, hair conditioners and hair coating shampoos. It is utilized as an antifoaming agent and cutting lubricant, as the precursor for the preparation of its sulfuric ester derivatives, which are used in detergents and wetting agents. It plays a vital role in various formulations for drug delivery.

    Solubility
    Miscible with alcohol and ether. Slightly miscible with carbon tetrachloride. Immiscible with water.

    Notes
    Incompatible with oxidizing agents and strong acids.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit