Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oleylamine, approximate C18-content 80-90%, ACROS Organics™

Click to view available options
Quantity:
5mL
100mL
1L
5L
112-90-3
C18H37N
267.501
QGLWBTPVKHMVHM-MDZDMXLPSA-N
9-octadecenylamine,9-octadecen-1-amine,cis-9-octadecenylamine,e-octadec-9-en-1-amine,1-amino-9-octadecene,9e octadec-9-enylamine,9-octadecenylamine 8ci,36505-83-6 hydrofluoride,3811-68-5 unspecified acetate
6258392
(E)-octadec-9-en-1-amine
CCCCCCCCC=CCCCCCCCCN
112-90-3
C18H37N
267.501
QGLWBTPVKHMVHM-MDZDMXLPSA-N
9-octadecenylamine,9-octadecen-1-amine,cis-9-octadecenylamine,e-octadec-9-en-1-amine,1-amino-9-octadecene,9e octadec-9-enylamine,9-octadecenylamine 8ci,36505-83-6 hydrofluoride,3811-68-5 unspecified acetate
6258392
(E)-octadec-9-en-1-amine
CCCCCCCCC=CCCCCCCCCN
Density 0.8100g/mL
Quantity 1L
Color Yellow
Refractive Index 1.4585 to 1.4625 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
Linear Formula CH3(CH2)7CH=CH(CH2)8NH2
Formula Weight 267.5
Physical Form Liquid After Melting
Percent Purity ≥96.0% (primary amine)
Chemical Name or Material Oleylamine
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger