Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Oleylethanolamide, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Lipid mediator and analog of anandamide (Cat. No. 1339) that is involved in peripheral regulation of feeding. Selective GPR55 agonist (EC50 values are 0.44, >30 and >30 μM at GPR55, CB1 and CB2 respectively) and PPARα agonist (EC50 = 120 nM). Induces satiety through activation of PPARα and is also a ceramidase inhibitor. Also endogenous agonist at the GPR119 receptor.
Specifications
Quantity | 10 mg |
Formula Weight | 325.53 |
Percent Purity | >99% |
Chemical Name or Material | Oleylethanolamide |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction