Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Olfactory Bioscience NOXO Odor Defense Pro™ Balm
No more retching, nausea, headaches or migraines
Manufacturer: Olfactory Bioscience NP0P75

Description
- Tin Size: 3 Ounces
- Great for detectives, nusing home visits and odor sensitivities
- Relief from the effects of odors
- Effective odor relief, smell only vanilla
- Safe and effective
- Made of 100% natural ingredients
- Apply balm over and around the nostril openings
- Special proprietary formulation of Phytonutrients and Phytochemicals taken from plants, flowers, roots and seeds
Specifications
Protection against odors | |
3 oz. | |
3 oz., 0.08L |
Vanilla | |
0.08L |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title