Learn More
Olfactory Marker Protein Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific OSR00038W
Catalog No. PIOSR00038W
Add to Cart
Description
Description
In IHC applications, Zamboni′s or 4% PFA fixatives are recommended.
Olfactory marker protein (OMP) is an abundant, 19-kDa, cytosolic protein that is almost exclusively expressed in mature, functioning, olfactory neurons but not in the neural precursor basal cells. The OMP gene structure and protein sequence are highly conserved between mouse, rat and human. Its tissue-specific expression in the receptor cells together with the results of the mouse knockout studies show that OMP represents a novel modulatory component of the odor detection/signal transduction cascade. May act as a modulator of the olfactory signal-transduction cascade. Interacts with BEX1 and BEX2. Uniquely associated with mature olfactory receptor neurons. Belongs to the olfactory marker protein family.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Olfactory Marker Protein
Polyclonal
Olfactory neuronal-specific protein; OMP
Synthetic peptide from hamster Olfactory Marker Protein.
RUO
Hamster
Antibody
IgG
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Rabbit
100 μL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Recommendations
Recommendations