Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OLFM3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549313

Catalog No. PA549313


Add to cart

Description

Description

Predicted to react with rat based on sequence homology.

Specifications

Specifications

OLFM3
Polyclonal
PBS with 0.09% sodium azide
Q96PB7, P63056
Noelin-3, Olfactomedin-3, Optimedin, OLFM3, NOE3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
118427, 229759
Western Blot
Unconjugated
OLFM3
Liquid
OLFM3
KLH conjugated synthetic peptide between 114-142 amino acids from the N-terminal region of human OLFM3.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit