Learn More
Aviva Systems Biology Corp OLFML1 Antibody - N-terminal region (ARP55873_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP55873_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OLFML1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human OLFML1. Alternate names: UNQ564ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61642