Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OLFML1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297287MFV610
Description
OLFML1 Polyclonal antibody specifically detects OLFML1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
OLFML1 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
MVAL564, olfactomedin-like 1, olfactomedin-like protein 1, UNQ564 | |
Produced in rabbits immunized with E. coli-derived Human OLFML1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
283298 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction