Learn More
Olfr554 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific OSR00202W
Catalog No. PIOSR00202W
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Reconstitute with 100 μL of distilled water. Centrifuge to remove any insoluble material.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Olfr554
Polyclonal
Olfr554
OLFR554
A synthetic peptide from mouse olfactory receptor family 25, subfamily 1
RUO
258322
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Immunohistochemistry, Western Blot
Unconjugated
Olfactory receptor MOR25-1, Olfr554
Rabbit
100 μL
Primary
Mouse
Antibody
IgG
Product Suggestions
Product Suggestions