Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Olfr611 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific OSR00197W
Description
Reconstitute with 100 µL of distilled water. Centrifuge to remove any insoluble material.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome.Specifications
Olfr611 | |
Polyclonal | |
Olfr611 | |
OLFR611 | |
Synthetic peptide from mouse olfactory receptor family 15, subfamily 1. | |
RUO | |
258722 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Olfactory receptor MOR15-1, MOR15-4, Olfr611 | |
Rabbit | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title