Learn More
Thermo Scientific Oligo(dT)18 Primers: PROMO
Thermo Scientific Oligo(dT)18 Primer is a synthetic single-stranded 18-mer oligonucleotide with 5'- and 3'-hydroxyl ends.
Manufacturer: Thermo Scientific SO131
Description
The primer is supplied as a ready-to-use, 20X concentrated aqueous solution for use in cDNA synthesis.Specifications
cDNA synthesis. | |
Oligo(dT)18 Primer | |
60μL (30μg) |