Learn More
Sklar InstrumentsSupplier Diversity Partner Olsen-Hegar Needle Holder, Sklar, 7 in., Blunt, Straight, Cross-Serrated with Groove, Finger Rings, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 202775
Olsen-Hegar Needle Holder, Sklar, 7 in., Blunt, Straight, Cross-Serrated with Groove, Finger Rings, Stainless Steel. Sklar's Olsen-Hegar Needle Holder is a ratcheted, finger ring instrument that also has built in scissors. It is used with intermediate weight sutures, commonly in plastic surgery procedures. This product is straight with serrated tips, and a central groove. It is 7 inches long.
Catalog No. 50-143-7489