Learn More
Sklar InstrumentsSupplier Diversity Partner Olsen-Hegar Needle Holder, Sklar, 4-3/4 in., Blunt, Straight, Smooth, Delicate, Finger Rings, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 202748
Olsen-Hegar Needle Holder, Sklar, 4-3/4 in., Blunt, Straight, Smooth, Delicate, Finger Rings, Stainless Steel. Sklar's Olsen-Hegar Needle Holder is a ratcheted, finger ring instrument that also has built in scissors. It is used with intermediate weight sutures, commonly in plastic surgery procedures. This product is straight with delicate, smooth tips, and an overall length of 4-3/4 inches.
Catalog No. 50-143-7486