Learn More
Tocris™ OLT 1177
NLRP3 inflammasome inhibitor; disrupts inflammasome formation
Manufacturer:  Tocris™ 6902/10
 View more versions of this product
Catalog No. 69-021-0
Add to cart
This item is not returnable. View return policy
Description
Description
Dapansutrile is a NLRP3 inflammasome inhibitor that disrupts inflammasome formation. Reduces LPS/ATP-induced IL-1β secretion in human and mouse macrophages in vitro, without affecting TNFα levels. Reduces LPS-induced inflammation in vivo. Also reduces joint swelling and markers of inflammation in synovial fluid in a mouse model of acute arthritis. Displays cardioprotective effects in a mouse model of cardiac ischemic reperfusion injury.
OLT1177® is a registered trademark of Olatec Therapeutics LLC in the United States and other countries.
Specifications
Specifications
133.17
NLRP3 inflammasome inhibitor; disrupts inflammasome formation
10 mg
C4H7NO2S
98%
Safety and Handling
Safety and Handling
Recommended Storage : Store at -20°C