Learn More
Tocris Bioscience™ Oltipraz
Nrf2 activator
$79.00 - $339.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
52-941-0 View Documents Tocris Bioscience™
5294/10
Each for $79.00
Add to cart
52-945-0 View Documents Tocris Bioscience™
5294/50
Each for $339.00
Add to cart
Description
Description
Nrf2 activator; activates Nrf, which binds to antioxidant response elements (AREs) and induces Phase-I and -II drug-metabolizing enzymes including CYP2B and NQO1. Also increases mRNA levels of multiple hepatic xenobiotic transporters. Elevates expression of genes encoding anti-oxidant and multidrug resistance-associated proteins. Displays chemopreventive activity.