EVOS™ Olympus Apochromat LWD Objective Lens
For demanding applications, especially capturing color images in white light
Manufacturer: EVOS™ AMEP4751
 View more versions of this product
Catalog No. 12-563-592
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) EVOS™ XL Core Imaging System, EVOS FL Color Imaging System, EVOS FL Auto Imaging System, EVOS XL Imaging System, EVOS FL Imaging System
    Product Type Olympus Apochromat LWD Objective Lens
    Numerical Aperture 0.08
    Magnification Power 2X
    Focus Distance 6.22mm
    • Has long working distance (LWD) objective that is optimized for imaging of slides, cell culture dishes and flasks, and microtiter plates
    • Compatible with EVOS™ Cell Imaging systems
    • Offers optical performance from visible light to near infrared light
    • Meets needs across spectrum of magnifications and optical specifications
  • Description & Specifications
Related Products
Product Content Feedback