EVOS™ Olympus Super-Apochromat LWD Objective Lens
Designed for demanding research applications
Manufacturer: EVOS™ AMEP4752
 View more versions of this product
Catalog No. 12-563-593
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) EVOS™ XL Core Imaging System, EVOS FL Color Imaging System, EVOS FL Auto Imaging System, EVOS XL Imaging System, EVOS FL Imaging System
    Product Type Super-Apochromat LWD Objective Lens
    Numerical Aperture 0.16
    Magnification Power 4X
    Focus Distance 13mm
    • Has long working distance (LWD) objective that is optimized for imaging of slides, cell culture dishes and flasks, and microtiter plates
    • Offers Apo correction from UV through IR ranges
    • Parfocal for both visible and IR imaging
    • Coverslip-corrected objective that is optimized for imaging through #1.5 coverslips
    • Compatible with EVOS Cell Imaging systems
    • Offers optical performance from visible light to near infrared light
    • Meets needs across spectrum of magnifications and optical specifications
  • Description & Specifications
Related Products
Product Content Feedback