Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ OMDM-2

Potent inhibitor of anandamide uptake

$67.40 - $282.50

Specifications

Assay Percent Range >99%
Formula Weight Observed MW: 431.66
Chemical Name or Material (9Z)-N-[1-((R)-4-Hydroxbenzyl)-2-hydroxyethyl]-9-octadecenamide
Molecular Formula C27H45NO3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
17-975-R View Documents Tocris Bioscience™
1797/5
5mg Each for $67.40
Add to cart
 
17-972-5 View Documents Tocris Bioscience™
1797/25
25mg Each for $282.50
Add to cart
 
Description

Description

Metabolically stable inhibitor of anandamide cellular uptake (Ki = 3 µM). Displays relatively low affinity for CB1 and CB2 receptors (Ki values are 5.1 and > 10 µM) and for vanilloid VR1 receptors (EC50 = 10 µM). Has minimal activity against FAAH (Ki > 50 µM). Active in vivo.
Specifications

Specifications

>99%
Observed MW: 431.66
(9Z)-N-[1-((R)-4-Hydroxbenzyl)-2-hydroxyethyl]-9-octadecenamide
C27H45NO3
Safety and Handling

Safety and Handling

Recommended Storage : Store at +4°C

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit