Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ Essentials Pipette Controller Replacement Filters

Catalog No. 03395127 Shop All Oakton Products
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
Sterility:
Non-sterile
Sterile
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Sterility
03-395-127 0.45 μm Sterile
03-395-125 0.2 μm Sterile
03-395-126 0.2 μm Non-sterile
03-395-128 0.45 μm Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 03-395-127 Supplier Cole-Parmer™ Essentials Supplier No. 2530339
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enhance and maintain your Omega Zen pipettors with these accessories

  • Compatible with Omega Zen pipette controllers
  • There are four filter options to choose from. All are made of a polypropylene housing with a hydrophobic PTFE membrane. The Omega Zen controllers accept either the 0.2 μm or the 0.45 μm pore size filters.

Warranty and Services

1 Year

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

Specifications

Autoclavable Not Autoclavable
Disposable Reusable
For Use With Omega Zen Pipettors
Material Hydrophobic PTFE
Model Omega Zen
Pore Size 0.45 μm
Sterility Sterile
Type Filter
Filter Type Hydrophobic PTFE
For Use With (Equipment) OmegaZen Pipette Controllers
Quantity 10
Warranty 1 Year
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.