Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OMG Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552530
Description
Immunogen sequence: VDTINSLSVV TQPKVTKIPK QYRTKETTFG ATLSKDTTFT STDKAFVPYP EDTSTETINS HEAAAATLTI HLQDGMVTNT SLTSSTKSSP TPMTLSITSG MPNNFSEMPQ QSTTLNLWRE ETTTNVKTPL PSVANAWKVN A Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 81%.
Oligodendrocyte myelin glycoprotein (OMG or OMGP) is a cell adhesion molecule contributing to the interactive process required for myelination in the central nervous system. It is also involved in axonogenesis, neuron projection regeneration, and regulation of collateral sprouting of intact axons in response to injury.
Specifications
OMG | |
Polyclonal | |
Unconjugated | |
OMG | |
oligodendrocyte myelin glycoprotein; oligodendrocyte-myelin glycoprotein; Omg; OMGP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4974 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.9 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P23515 | |
OMG | |
Recombinant protein corresponding to Human OMG. Recombinant protein control fragment (Product #RP-89911). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction