Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Omni International Fine Saw Tooth Generator Probe
Saw Teeth add extra cutting action and stator design allows fine to medium samples to pass through the processing windows.
Supplier: Omni International G795ST
Description
Comes with PTFE upper and lower bearing. The entire probe can be autoclaved, or easily disassembled for cleaning. Optional Stainless steel upper bearings extend bearing life but are not autoclavable.Specifications
Fine Sawtooth Generator Probe | |
7 mm dia. x 95 mm L | |
Liquid/liquid and fibrous tissue samples | |
PTFE upper and lower bearing |
5 mm | |
Stainless Steel | |
Omni Mixer |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction