Learn More
Omni International General Laboratory Homogenizer
High power motor can handle tough tissue samples
Manufacturer:  Omni International IncSupplier Diversity Partner GLH115
 View more versions of this product
Catalog No. 15-340-108
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
200μL to 10L
7cm
Ideal for soft and hard tissue homogenization, cellular, biological, cancer research
<72dBA
115V
2.75 in.
General Laboratory Homogenizer
5, 7, 10, 20, 32, 35mm dia. generator probes
5000 to 28000rpm