Learn More
Omni International Omni Prep™ Multisample Homogenizer Available on GSA/VA Contract for Federal Government customers only.
Homogenize six samples per cycle—up to 250 samples per hour
Manufacturer:  Omni International 06021
Catalog No. 15-340-451
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Description
Description
Modular system† allows one group of samples to be prepared while another group is being processed. Processes samples rapidly and efficiently in a variety of tube sizes, including 1.5/2, 15, and 50mL conical tubes.
  • Unit works exclusively with Omni Tip™ plastic probes
  • Omni Tip probes are disposable to eliminate cross-contamination; may also be reused and autoclaved up to seven times
  • Programmable time and speed—stores up to three programs
  • Most samples processed in under one minute
  • Movable rack is ideal for processing samples that require probe mobility within the sample
  • Allows processing in open or sealed vessels
  • Fan-driven positive airflow can be exhausted to a HEPA filter or fume hood
  • Ice tray (sold separately) keeps sensitive samples cold during processing
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Certifications
CE marked
Warranty and Services
One year
Specifications
Specifications
115/230V
0.25 to 30mL
Up to 10 min.
150W
9.53kg
115/230V 50/60Hz
500 to 30000rpm
Multi-Sample Homogenizer
0.008 to 1.01 oz.
21 lb.
15.74 x 7.87 x 11.75 in. (40 x 20 x 30cm)