Learn More
Kinesis™ Omnifit™ Benchmark™ Low-Pressure Chromatography Column Assemblies
Designed to suit the majority of standard laboratory chromatography applications, including protein purification
Manufacturer:  Kinesis™ 006BCC0640FF
 View more versions of this product
Catalog No. 14-500-674
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • For aqueous systems
  • Economical modular columns for low pressure or gravity feed, providing even distribution of eluent to the medium bed, with or without flow adapters
  • Ready-assembled column includes PTFE endpieces and 25µm PE frits
Specifications
Specifications
25μm
400mm
Borosilicate Glass
6.6mm
Documents
Accessories