Learn More
Diba Omnifit Cap Adapters
Adapters replace the cap on any Omnifit cap connector or valve
$47.50 - $50.80
Description
- Enable quick push-on connections for softwall tubing or Luer connectors
- Autoclavable
Specifications
Cap Adapter |