Learn More
Kinesis™ Omnifit™ Chromatography Column Replacement Endpieces: Adjustable
Manufacturer:  Kinesis™ 006BAE06
 View more versions of this product
Catalog No. 14-500-687
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Specifications
PE frit
0.66 cm
900 psi
4°C to 60°C
0.25 in. to 28 flat bottom
Adjustable endpiece
Adjustable Length
0.25 in.
BenchMark Chromatography Columns to Distribute Sample Evenly Across Bed Surface
39.2°F to 140°F
25 µm
900 psi
1 to 14
PTFE