Learn More
Kinesis Omnifit Chromatography Column Replacement Endpieces: Fixed
Manufacturer: Kinesis 006BFE0350
Specifications
0.11 in. | |
3 mm | |
50 µm | |
PTFE |
0.11 in. | |
BenchMark Columns Used in Chromatography | |
Fixed Length |