Learn More
Kinesis Omnifit Glass Columns for Low/Medium Pressure Liquid Chromatography
Standard aqueous system with a borosilicate glass chromatographic tube and a PTFE plunger
Specifications
Endcapped | Yes |
---|---|
Material | Borosilicate Glass |
Requires: Replacement glass columns, fixed and adjustable endpieces, frits, and O-rings are available separately; accessories such as frit insertion and removal tools, packing sleeves, column jackets, silicone seals, septum injectors, and bleed valves are also available separatelycontact your Fisher Scientific Customer Service Representative for details.
Catalog Number | Column Capacity | Max. Pressure | Connector Types | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Column Capacity | Max. Pressure | Connector Types | Price | Quantity & Availability | ||||
14-500-645
|
5.5 mL | 600 psi | Two fixed endpieces |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
Description
- Robust and easy to use
- Biocompatible
- Choice of columns with fixed or adjustable height; columns with an adjustable endpiece have a maximum height of 80mm (designed with fine adjustment control to minimize the risk of disturbing the packed bed)
- For a solvent-resistant system able to withstand the harshest laboratory chemicals, simply replace the 25µm polyethylene frit in each endpiece with the PTFE frit in the included frit kit
Columns with 3.0mm I.D. (bore size) are only available with fixed endpieces on both ends and do not include frit kits.
Specifications
Yes | |
Borosilicate Glass |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.