Learn More
Omnifit™ Glass Columns for Low/Medium Pressure Liquid Chromatography: Replacement Columns
Standard aqueous system with a borosilicate glass chromatographic tube and a PTFE plunger
Manufacturer:  OmnifitSupplier Diversity Partner 006BCC0605AF
 View more versions of this product
Catalog No. 14-500-632
  Check Availability
Add to cart
Requires : Replacement glass columns, fixed and adjustable endpieces, frits, and O-rings are available separately; accessories such as frit insertion and removal tools, packing sleeves, column jackets, silicone seals, septum injectors, and bleed valves are also available separately—contact your Fisher Scientific Customer Service Representative for details.
Specifications Description & Specifications
Specifications
4 to 20°C
Empty Chromatography Column
900psi
1 to 4
50mm
6.6mm
Low/Medium pressure LC
Yes
25μm
0.7mL
Borosilicate Glass
Ready assembled in FF, AF or AA configuration of precision bore Borosilicate Glass column, 2 endpieces with 25μm Polyethylene frits, frit kit comprising of 2 x 30 and 10μm PTFE frits, 2 x 10μm Polyethylene frits, fittings kit comprising of 2 and 0.5m x 0.062 in. O.D. PTFE tubing with 0.25 in. 28 UNF fitting, two M6 endcaps, FKM/FPM O ring, Acetal adjusting nut and retaining cap, Glass filled Polypropylene connection cap and fitting nuts
Accessories