Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diba Omnifit™ Replacement Endpieces for EZ SolventPlus™ Columns
Select from fixed or adjustable Omnifit™ Replacement Endpieces for EZ SolventPlus™ Columns
Supplier: Diba 1194271
Specifications
Liquid Chromatography Column | |
Fixed | |
Omnifit™ EZ SolventPlus™ EZ SolventPlus Fixed Column Endpiece, 50mm; 1/ea |
50 mm | |
Omnifit™ EZ SolventPlus™ Columns | |
PTFE |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction