Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Omnifit™ Replacement Glass Tubes for EZ and EZ SolventPlus™ Columns

Mix and match your Omnifit™ Replacement Glass Tubes for EZ and EZ SolventPlus™ Column lengths while using your existing endpieces for ultimate flexibility

Manufacturer:  OmnifitSupplier Diversity Partner 006ERG1033

 View more versions of this product

Catalog No. 14-533-210


Add to cart

Description

Description

Use a different length tube with your existing endpieces to meet your specific requirements.
Specifications

Specifications

Omnifit™ EZ Replacement Glass, 10 x 330mm; 1/ea
Omnifit™ EZ and EZ SolventPlus™ Columns
330mm
Liquid Chromatography (LC)
10mm
Glass
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit