Omnifit™ SolventPlus™ Chromatography Columns and Replacement Parts
Designed for use with organic solvents but equally compatible with aqueous solvent systems
Manufacturer: OmnifitSupplier Diversity Partner 006SCC0625FF
 View more versions of this product
Catalog No. 14-500-669
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    For Use With Omnifit SolventPlus chromatography columns
    Diameter (Metric) Inner 6.6mm
    Diameter (Metric) Outer 6.6mm
    Includes Glass column assembly with PTFE endpiece, frit; 2 fixed endpieces
    Length (Metric) 250mm
    Type SolventPlus chromatography column
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback